Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174291.2 | NP_776716.1 | 2 - 3 | 277 | 72 | |
GenBank mRNA | L27070.1 | - | 2 - 3 | 277 | 72 | |
M16895.1 | - | 2 - 3 | 277 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Bos taurus Probe spans exons |