Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174348.2 | NP_776773.1 | 4 - 5 | 1474 | 95 | |
GenBank mRNA | EU277736.1 | - | 4 - 5 | 921 | 95 | |
U65789.1 | - | 4 - 5 | 1474 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |