Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174358.2 | NP_776783.1 | 1 - 2 | 253 | 67 | |
GenBank mRNA | BC133546.1 | - | 1 - 2 | 221 | 67 | |
M20818.1 | - | 1 - 2 | 253 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |