Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174379.2 | NP_776804.1 | 4 - 5 | 652 | 60 | |
GenBank mRNA | AB098757.1 | - | 2 - 3 | 328 | 60 | |
AB098775.1 | - | 2 - 3 | 379 | 60 | ||
BC102490.1 | - | 5 - 6 | 702 | 60 | ||
JF979129.1 | - | 4 - 5 | 624 | 60 | ||
M64923.1 | - | 4 - 5 | 652 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |