Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPCML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174407.2 | NP_776832.1 | 5 - 6 | 1565 | 77 | |
GenBank mRNA | X12672.1 | - | 1565 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |