Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE6D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174420.2 | NP_776845.1 | 3 - 4 | 424 | 138 | |
GenBank mRNA | U65073.1 | - | 3 - 4 | 424 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |