Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174433.2 | NP_776858.1 | 1 - 2 | 338 | 93 | |
GenBank mRNA | AF305563.1 | - | 1 - 2 | 338 | 93 | |
BC109824.1 | - | 1 - 2 | 305 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |