Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174465.2 | NP_776890.1 | 3 - 4 | 398 | 114 | |
GenBank mRNA | AF221669.1 | - | 3 - 4 | 398 | 114 | |
BC102089.1 | - | 3 - 4 | 402 | 114 | ||
BC150010.1 | - | 3 - 4 | 427 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |