Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174524.2 | NP_776949.1 | 3 - 4 | 349 | 94 | |
GenBank mRNA | M60329.1 | - | 3 - 4 | 349 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |