Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174539.2 | NP_776964.1 | 4 - 5 | 725 | 86 | |
GenBank mRNA | BC113215.1 | - | 4 - 5 | 713 | 86 | |
EU151552.1 | - | 1 - 2 | 199 | 86 | ||
X75671.1 | - | 725 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |