Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP11B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174638.3 | NP_777063.2 | 3 - 4 | 602 | 127 | |
GenBank mRNA | AH003250.2 | - | 3 - 4 | 600 | 127 | |
D00185.1 | - | 602 | 127 |
Gene Symbol: LOC787647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | D00361.1 | - | 604 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |