Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAFAH1B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174664.2 | NP_777089.1 | 2 - 3 | 485 | 87 | |
GenBank mRNA | AB098963.1 | - | 2 - 3 | 158 | 87 | |
BC103452.1 | - | 2 - 3 | 171 | 87 | ||
D49678.1 | - | 2 - 3 | 485 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |