Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174767.2 | NP_777192.1 | 1 - 2 | 462 | 77 | |
GenBank mRNA | AF251032.1 | - | 1 - 2 | 462 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |