Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPING1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174821.2 | NP_777246.1 | 4 - 5 | 810 | 79 | |
GenBank mRNA | U30332.1 | - | 4 - 5 | 810 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |