Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176664.2 | NP_788837.1 | 2 - 3 | 220 | 67 | |
GenBank mRNA | X02509.1 | - | 220 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |