Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177488.3 | NP_803454.2 | 7 - 8 | 804 | 97 | |
GenBank mRNA | BC102939.1 | - | 6 - 7 | 704 | 97 | |
EV793129.1 | - | 4 - 5 | 379 | 97 | ||
M54942.1 | - | 8 - 9 | 800 | 97 | ||
X56395.1 | - | 800 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |