Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLT3LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181030.2 | NP_851373.1 | 6 - 7 | 674 | 87 | |
GenBank mRNA | AB051841.1 | - | 6 - 7 | 674 | 87 | |
AF282985.1 | - | 6 - 7 | 674 | 87 | ||
AF282986.1 | - | 6 - 7 | 620 | 87 | ||
BC133533.1 | - | 7 - 8 | 696 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |