Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205777.2 | NP_991346.2 | 2 - 3 | 194 | 81 | |
NM_205818.2 | NP_991387.2 | 2 - 3 | 241 | 81 | ||
GenBank mRNA | AY911378.1 | - | 2 - 3 | 165 | 81 | |
BC126769.1 | - | 2 - 3 | 173 | 81 | ||
BC148015.1 | - | 2 - 3 | 208 | 81 | ||
CB454302.1 | - | 3 - 4 | 540 | 81 | ||
CB455704.1 | - | 2 - 3 | 203 | 81 | ||
CB463455.1 | - | 1 - 2 | 141 | 81 | ||
CR454063.1 | - | 2 - 3 | 222 | 81 | ||
EE337437.1 | - | 2 - 3 | 242 | 81 | ||
X92956.1 | - | 180 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |