Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205794.1 | NP_991363.1 | 2 - 3 | 218 | 104 | |
GenBank mRNA | AY533678.1 | - | 2 - 3 | 218 | 104 | |
BC151491.1 | - | 2 - 3 | 326 | 104 | ||
BT030588.1 | - | 2 - 3 | 279 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |