Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAPTM4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205802.2 | NP_991371.2 | 3 - 4 | 526 | 72 | |
GenBank mRNA | AF276819.1 | - | 3 - 4 | 477 | 72 | |
BC149605.1 | - | 3 - 4 | 524 | 72 | ||
EH143146.1 | - | 3 - 4 | 524 | 72 | ||
EV638847.1 | - | 3 - 4 | 235 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |