Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC149891.1 | - | 1 - 2 | 94 | 66 | |
BM432047.1 | - | 1 - 2 | 38 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |