Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013603.1 | NP_001013621.1 | 4 - 5 | 545 | 94 | |
GenBank mRNA | BC103453.1 | - | 4 - 5 | 572 | 94 | |
BT021871.1 | - | 4 - 5 | 545 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |