Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15H11orf16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014846.1 | NP_001014846.1 | 4 - 5 | 738 | 92 | |
GenBank mRNA | BC102543.1 | - | 5 - 6 | 809 | 92 | |
BT021678.1 | - | 4 - 5 | 738 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |