Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014874.1 | NP_001014874.1 | 4 - 5 | 527 | 72 | |
GenBank mRNA | BT021695.1 | - | 4 - 5 | 527 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |