Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014945.2 | NP_001014945.1 | 1 - 2 | 31 | 79 | |
GenBank mRNA | AY911382.1 | - | 1 - 2 | 32 | 79 | |
BC105345.1 | - | 1 - 2 | 32 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |