Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014960.1 | NP_001014960.1 | 4 - 5 | 405 | 80 | |
GenBank mRNA | BC114889.1 | - | 3 - 4 | 308 | 80 | |
BT021537.1 | - | 4 - 5 | 405 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |