Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015598.1 | NP_001015598.1 | 1 - 2 | 1200 | 58 | |
GenBank mRNA | BC146253.1 | - | 1 - 2 | 1204 | 58 | |
BT021600.1 | - | 1 - 2 | 1200 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |