Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015615.1 | NP_001015615.1 | 2 - 3 | 285 | 123 | |
GenBank mRNA | BC102367.1 | - | 2 - 3 | 287 | 123 | |
BT020855.1 | - | 2 - 3 | 260 | 123 | ||
BT020920.1 | - | 2 - 3 | 285 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |