Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015674.1 | NP_001015674.1 | 4 - 5 | 626 | 91 | |
GenBank mRNA | BT021859.1 | - | 4 - 5 | 626 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |