Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZGPAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024514.2 | NP_001019685.2 | 3 - 4 | 857 | 90 | |
GenBank mRNA | BC118131.1 | - | 3 - 4 | 849 | 90 | |
BT021717.1 | - | 2 - 3 | 938 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |