Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024540.1 | NP_001019711.1 | 4 - 5 | 438 | 137 | |
GenBank mRNA | BC109840.1 | - | 4 - 5 | 440 | 137 | |
BT021522.1 | - | 4 - 5 | 438 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |