Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF1AX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025329.2 | NP_001020500.1 | 1 - 2 | 179 | 98 | |
GenBank mRNA | AY911362.1 | - | 1 - 2 | 180 | 98 | |
BC109650.1 | - | 1 - 2 | 238 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |