Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF1AX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025329.2 | NP_001020500.1 | 2 - 3 | 267 | 75 | |
GenBank mRNA | AY911362.1 | - | 2 - 3 | 268 | 75 | |
BC109650.1 | - | 2 - 3 | 326 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |