Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031852.1 | NP_001027022.1 | 4 - 5 | 499 | 70 | |
GenBank mRNA | AY937382.1 | - | 3 - 4 | 358 | 70 | |
AY937383.1 | - | 4 - 5 | 499 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |