Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIKESHI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034226.2 | NP_001029398.1 | 3 - 4 | 625 | 88 | |
GenBank mRNA | AY911348.1 | - | 3 - 4 | 626 | 88 | |
BC103227.1 | - | 3 - 4 | 570 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |