Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTRH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034519.1 | NP_001029691.1 | 1 - 2 | 227 | 114 | |
GenBank mRNA | BT021191.1 | - | 1 - 2 | 227 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |