Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034677.1 | NP_001029849.1 | 5 - 6 | 686 | 88 | |
GenBank mRNA | BC102645.1 | - | 5 - 6 | 686 | 88 | |
BT030497.1 | - | 5 - 6 | 651 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |