Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034742.1 | NP_001029914.1 | 4 - 5 | 508 | 85 | |
GenBank mRNA | BT020918.1 | - | 4 - 5 | 508 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |