Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035370.2 | NP_001030447.2 | 5 - 6 | 1058 | 113 | |
GenBank mRNA | BC120240.1 | - | 4 - 5 | 906 | 113 | |
BT020815.1 | - | 5 - 6 | 1058 | 113 | ||
BT020938.1 | - | 4 - 5 | 832 | 113 | ||
BT026296.1 | - | 5 - 6 | 1048 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |