Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BT021865.1 | - | 2 - 3 | 245 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |