Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035455.1 | NP_001030532.1 | 5 - 6 | 780 | 95 | |
GenBank mRNA | BC146213.1 | - | 5 - 6 | 801 | 95 | |
BT020979.1 | - | 5 - 6 | 780 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |