Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075790.1 | NP_001069258.1 | 3 - 4 | 213 | 72 | |
GenBank mRNA | BC142193.1 | - | 3 - 4 | 213 | 72 | |
BT021737.1 | - | 3 - 4 | 213 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |