Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077003.1 | NP_001070471.1 | 5 - 6 | 865 | 79 | |
GenBank mRNA | BC103238.1 | - | 5 - 6 | 865 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |