Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078126.2 | NP_001071594.1 | 3 - 4 | 358 | 77 | |
GenBank mRNA | BC103100.1 | - | 3 - 4 | 360 | 77 | |
BC149575.1 | - | 4 - 5 | 808 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |