Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174234.1 | NP_776659.1 | 5 - 6 | 783 | 69 | |
GenBank mRNA | AF296410.1 | - | 5 - 6 | 783 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Bos taurus Probe spans exons |