Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174274.1 | NP_776699.1 | 1 - 2 | 95 | 120 | |
GenBank mRNA | AY027881.1 | - | 1 - 2 | 134 | 120 | |
AY027882.1 | - | 1 - 2 | 134 | 120 | ||
BT025444.1 | - | 1 - 2 | 89 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |