Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VEGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174488.2 | NP_776913.1 | 2 - 3 | 486 | 106 | |
GenBank mRNA | AB004275.1 | - | 2 - 3 | 486 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |