Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174558.2 | NP_776983.1 | 4 - 5 | 742 | 121 | |
GenBank mRNA | AF479760.1 | - | 4 - 5 | 742 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |