Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174678.3 | NP_777103.1 | 1 - 2 | 491 | 107 | |
GenBank mRNA | AF453510.1 | - | 1 - 2 | 492 | 107 | |
AF453511.1 | - | 1 - 2 | 492 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |