Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFDN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174732.2 | NP_777157.1 | 2 - 3 | 180 | 70 | |
GenBank mRNA | AF520958.1 | - | 2 - 3 | 180 | 70 | |
BC102252.1 | - | 2 - 3 | 188 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |